Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNase-free Tips, 1,000 μL size

Catalog No. AM12660
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
AM12660 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM12660 Supplier Invitrogen™ Supplier No. AM12660TS
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Provided in ten (10) racks containing 100 tips each. Invitrogen™ Ambion™ RNase-free Tips, 1000μL size are subjected to rigorous testing for RNase contamination and are certified to be RNase- and DNase-free.

Invitrogen™ Tips are compatible with Eppendorf™, Finnpipette, Oxford Benchmate™, Pipetman™, Proline™, Rainin, Socorex, and other brands of pipettors.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Sterile
Product Type RNase-free Tips
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
For Use With (Equipment) Universal Fit
Product Line Ambion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.