Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Spin Columns and Tubes

Catalog No. AM10065
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM10065 100 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM10065 Supplier Invitrogen™ Supplier No. AM10065
Add to Cart
Add to Cart

Includes

30 columns

For rapid elution of DNA from agarose gel slices

Simply excise a band from the gel, place it in the spin column, and centrifuge for five minutes. DNA is forced through the column into the tube below, while agarose is left behind. DNA is ready for subsequent manipulations such as restriction enzyme digestion or ligation.

  • No electroelution is needed; columns only require microcentrifuge
  • Certified DNase- and RNase-free

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Filtered Filtered
For Use With (Application) DNA Extraction from Agarose Gels
Format Tube, Spin Column
Tip Style Universal Fit
Quantity 100 units
Product Type Spin Columns and Tubes
Content And Storage Store tubes at room temperature.
Shipping Condition Room Temperature
Disposable Single-use
For Use With (Equipment) Microcentrifuge
Product Line Invitrogen
Show More Show Less
Do you offer an alternative for the discontinued NucAway Spin Columns?

Yes, we recommend Spin Columns and Tubes (Cat. No. AM10065) (https://www.thermofisher.com/order/catalog/product/AM10065).

Do you offer an alternative to the discontinued NucAway Spin Columns (Cat. No. AM10070)?

We recommend using Spin Columns and Tubes (Cat. No. AM10065): https://www.thermofisher.com/order/catalog/product/AM10065

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.