Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TE, pH 7.0, RNase-free

Catalog No. p-4926936
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
10 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9860 10 x 1 mL
AM9861 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM9860 Supplier Invitrogen™ Supplier No. AM9860
Only null left
Add to Cart
Edge
Add to Cart

Invitrogen TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0 is in nuclease-free, ultrapure water.

Invitrogen TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0 is in nuclease-free, ultrapure water. The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. TE Buffer is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

pH 7.0
Physical Form Liquid
Product Line Ambion
Quantity 10 x 1 mL
Packaging Bottle

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.