Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tris (1 M), pH 7.0, RNase-free

Catalog No. p-4926941
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9850G 100 mL
AM9851 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM9850G Supplier Invitrogen™ Supplier No. AM9850G
Only null left
Add to Cart
Edge
Add to Cart

Invitrogen Molecular biology grade, 1M Tris, pH 7.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use.

Invitrogen Molecular biology grade, 1M Tris, pH 7.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 M
pH 7
Physical Form Liquid
Product Line Ambion
Quantity 100 mL
Formulation 1,3-Propanediol, 2-Amino-2-(Hydroxymethyl)-,Hydrochloride
Grade Molecular Biology
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.