Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UTP Soln. (10 mM)

Catalog No. AM8140G
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8140G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8140G Supplier Invitrogen™ Supplier No. AM8140G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits and is supplied in one tube containing 100 μL.

This Invitrogen ribonucleotide triphosphate (rNTP) solution is a component of the MAXIscript™ Kits (Cat. No. AM1312, AM1316, AM1314, AM1326, AM1324, AM1318, AM1322, AM1320, AM1310, and AM1308) and is supplied in one tube containing 100 μL. Each is subjected to RNase testing as well as a functional assay to ensure high levels of incorporation.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Concentration 10 mM
For Use With (Application) In Vitro transcription
Label or Dye Unlabeled
Product Line Ambion
Quantity 100 μL
Shipping Condition Dry Ice
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.