Learn More
Aviva Systems Biology Corp AMBP Antibody - N-terminal region (ARP59163_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59163_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMBP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human AMBP. Alternate names: A1M, EDC1, HCP, HI30, IATIL, ITI, ITIL, ITILC, UTIApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-286-601