Learn More
Uline Amber Boston Round Glass Bottles - 1 oz
Catalog No. NC3455062
Encompass
Store and pour chemicals and medicines.Narrow mouth for precision pouring.Safe storage for hydrogen peroxide, sulfuric acid and light-sensitive chemicals.FDA compliant.Phenolic black caps included.
Catalog No. NC3455062 Supplier Uline Supplier No. S20888
Store and pour chemicals and medicines.Narrow mouth for precision pouring.Safe storage for hydrogen peroxide, sulfuric acid and light-sensitive chemicals.FDA compliant.Phenolic black caps included.