Learn More
Analytical Sales & Services AMBR SCRW VL & CP PTFE/SIL LIN
Catalog No. NC1936491
Encompass
Unassembled Precision MSQ Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Screw Vials and 9mm Screw Caps with Pre-Slit MSQ PTFE/Silicone Liners. 1000/CS ANALYTICAL SALES & SERVICE mfg# 99005ASL-CASE
Catalog No. NC1936491 Supplier Analytical Sales & Services Supplier No. 99005ASLCASE
Unassembled Precision MSQ Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Screw Vials and 9mm Screw Caps with Pre-Slit MSQ PTFE/Silicone Liners. 1000/CS ANALYTICAL SALES & SERVICE mfg# 99005ASL-CASE