Learn More
Apexbio Technology LLC Ambrisentan 177036-94-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062152
Encompass_Preferred
Ambrisentan (CAS 177036-94-1) is an orally active selective endothelin receptor antagonist specifically targeting ETA receptors It exhibits strong selectivity towards ETA over ETB receptor subtypes shown by Ki values of approximately 1 nM for recombinant ETA receptors expressed in vitro versus 195 nM for ETB In recombinant human receptor assays using intact cells this selectivity is enhanced with reported Ki values of 0 63 nM (ETA) and 48 7 nM (ETB) Pharmacologically ambrisentan s selective ETA blockade preserves the ETB receptor s intrinsic vasodilatory and endothelin clearance functions making it useful in research involving pulmonary arterial hypertension cerebrovascular conditions and ischemia-reperfusion injury
Catalog No. 50-000-62152 Supplier Apexbio Technology LLC Supplier No. B20755.1
Ambrisentan (CAS 177036-94-1) is an orally active selective endothelin receptor antagonist specifically targeting ETA receptors It exhibits strong selectivity towards ETA over ETB receptor subtypes shown by Ki values of approximately 1 nM for recombinant ETA receptors expressed in vitro versus 195 nM for ETB In recombinant human receptor assays using intact cells this selectivity is enhanced with reported Ki values of 0 63 nM (ETA) and 48 7 nM (ETB) Pharmacologically ambrisentan s selective ETA blockade preserves the ETB receptor s intrinsic vasodilatory and endothelin clearance functions making it useful in research involving pulmonary arterial hypertension cerebrovascular conditions and ischemia-reperfusion injury