Learn More
Apexbio Technology LLC Ambrisentan, 5mg. Cas: 177036-94-1 MFCD: MFCD08672619. ETA-receptor antagonist
SDP
Supplier:  Apexbio Technology LLC B20755
Encompass_Preferred
Ambrisentan is a selective antagonist of ETA-receptor with Ki value of 1nM [1].Ambrisentan is an orally active diphenyl propionic acid derivative. It is usually used to treat for pulmonary arterial hypertension. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5304