Learn More
Cayman Chemical Ambrisentan-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1189479-60-4, 1 mg
SDP
Catalog No. 502910084
Encompass_Preferred
An internal standard for the quantification of ambrisentan by GC- or LC-MS
Catalog No. 50-291-0084 Supplier Cayman Chemical Supplier No. 355581 MG
An internal standard for the quantification of ambrisentan by GC- or LC-MS