Learn More
Medchemexpress LLC Ambroxol-d5 (hydrochloride) | 2741380-71-0 | 99.6% | 500 UG
SDP
Catalog No. 5000413805
Encompass_Preferred
Ambroxol-d5 (hydrochloride) is the deuterium-labeled Ambroxol hydrochloride. Ambroxol hydrochloride, an active metabolite of the proagent Bromhexine, has potent expectorant effects. It acts as a glucocerebrosidase (GCase) chaperone, increasing glucocerebrosidase activity and inducing lung autophagy. It has potential for Parkinson's disease and neuronopathic Gaucher disease research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13805 Supplier Medchemexpress LLC Supplier No. HYB1039AS500UG
Ambroxol-d5 (hydrochloride) is the deuterium-labeled Ambroxol hydrochloride. Ambroxol hydrochloride, an active metabolite of the proagent Bromhexine, has potent expectorant effects. It acts as a glucocerebrosidase (GCase) chaperone, increasing glucocerebrosidase activity and inducing lung autophagy. It has potential for Parkinson's disease and neuronopathic Gaucher disease research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.