Learn More
Medchemexpress LLC Ambroxol (hydrochloride) | 23828-92-4 | 100.0% | 10 MM 1 ML
SDP
Catalog No. 5000413881
Encompass_Preferred
Ambroxol hydrochloride is an active metabolite of Bromhexine with potent expectorant effects. It functions as a glucocerebrosidase (GCase) chaperone, enhancing glucocerebrosidase activity. This compound also induces lung autophagy and is a potential area of research for Parkinson's disease and neuronopathic Gaucher disease.
  • Active metabolite of Bromhexine
  • Potent expectorant effects
  • Acts as a glucocerebrosidase (GCase) chaperone
  • Increases glucocerebrosidase activity
  • Induces lung autophagy
  • Potential for research in Parkinson's disease
  • Potential for research in neuronopathic Gaucher disease
  • High purity (99.96%)
  • White to off-white solid appearance
Catalog No. 50-004-13881 Supplier Medchemexpress LLC Supplier No. HYB1039A10MM/1ML
Ambroxol hydrochloride is an active metabolite of Bromhexine with potent expectorant effects. It functions as a glucocerebrosidase (GCase) chaperone, enhancing glucocerebrosidase activity. This compound also induces lung autophagy and is a potential area of research for Parkinson's disease and neuronopathic Gaucher disease.
  • Active metabolite of Bromhexine
  • Potent expectorant effects
  • Acts as a glucocerebrosidase (GCase) chaperone
  • Increases glucocerebrosidase activity
  • Induces lung autophagy
  • Potential for research in Parkinson's disease
  • Potential for research in neuronopathic Gaucher disease
  • High purity (99.96%)
  • White to off-white solid appearance