Learn More
Medchemexpress LLC Ambroxol hydrochloride | 23828-92-4 | 99.96% | 100 MG
SDP
Catalog No. 5000411876
Encompass_Preferred
Ambroxol hydrochloride (NA-872 hydrochloride) is an active metabolite of Bromhexine, known for its potent expectorant effects. It acts as a glucocerebrosidase (GCase) chaperone, increasing glucocerebrosidase activity. This compound also induces lung autophagy and shows potential for research in Parkinson's disease and neuronopathic Gaucher disease.
  • Potent expectorant effects.
  • Increases glucocerebrosidase activity.
  • Induces lung autophagy.
  • Potential for Parkinson's disease research.
  • Potential for neuronopathic Gaucher disease research.
Catalog No. 50-004-11876 Supplier Medchemexpress LLC Supplier No. HYB1039A100MG
Ambroxol hydrochloride (NA-872 hydrochloride) is an active metabolite of Bromhexine, known for its potent expectorant effects. It acts as a glucocerebrosidase (GCase) chaperone, increasing glucocerebrosidase activity. This compound also induces lung autophagy and shows potential for research in Parkinson's disease and neuronopathic Gaucher disease.
  • Potent expectorant effects.
  • Increases glucocerebrosidase activity.
  • Induces lung autophagy.
  • Potential for Parkinson's disease research.
  • Potential for neuronopathic Gaucher disease research.