Learn More
Medchemexpress LLC Ambroxol (hydrochloride) | 23828-92-4 | 99.96% | 500 MG
SDP
Catalog No. 5000413854
Encompass_Preferred
Ambroxol hydrochloride, an active metabolite of Bromhexine, possesses potent expectorant effects. It acts as a glucocerebrosidase (GCase) chaperone, increasing glucocerebrosidase activity. This compound also induces lung autophagy and shows potential for research in Parkinson's disease and neuronopathic Gaucher disease.
  • Potent expectorant effects
  • Increases glucocerebrosidase activity
  • Induces lung autophagy
  • Potential for research in Parkinson's disease
  • Potential for research in neuronopathic Gaucher disease
Catalog No. 50-004-13854 Supplier Medchemexpress LLC Supplier No. HYB1039A500MG
Ambroxol hydrochloride, an active metabolite of Bromhexine, possesses potent expectorant effects. It acts as a glucocerebrosidase (GCase) chaperone, increasing glucocerebrosidase activity. This compound also induces lung autophagy and shows potential for research in Parkinson's disease and neuronopathic Gaucher disease.
  • Potent expectorant effects
  • Increases glucocerebrosidase activity
  • Induces lung autophagy
  • Potential for research in Parkinson's disease
  • Potential for research in neuronopathic Gaucher disease