Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ambroxol hydrochloride | 23828-92-4 | MFCD00078932 | 99.9% | 414.56 g/mol | C13H19Br2ClN2O | 10 MG
SDP

Catalog No. 5000298211
Encompass_Preferred

Ambroxol hydrochloride analytical standard supplied as a white to off-white solid reference material for research and analytical applications. The material is provided at high stated purity for assay development, method validation, and quality control in laboratory settings.

  • High purity: 99.91% (analytical standard)
  • CAS number: 23828-92-4
  • Molecular weight: 414.56 g/mol
  • Appearance: white to off-white solid suitable for reference use
  • Supplied in small packaged amounts appropriate for lab testing, e.g., 10 MG
  • Intended uses: reference standard, analytical calibration, and QC applications

Catalog No. 50-002-98211 Supplier Medchemexpress LLC Supplier No. HYB1039AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ambroxol hydrochloride analytical standard supplied as a white to off-white solid reference material for research and analytical applications. The material is provided at high stated purity for assay development, method validation, and quality control in laboratory settings.

  • High purity: 99.91% (analytical standard)
  • CAS number: 23828-92-4
  • Molecular weight: 414.56 g/mol
  • Appearance: white to off-white solid suitable for reference use
  • Supplied in small packaged amounts appropriate for lab testing, e.g., 10 MG
  • Intended uses: reference standard, analytical calibration, and QC applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.