Learn More
Medchemexpress LLC Ambroxol hydrochloride (Standard) | 23828-92-4 | 99.9% | 100 MG
SDP
Catalog No. 5000412440
Encompass_Preferred
Ambroxol hydrochloride (Standard) is an analytical standard and an active metabolite of Bromhexine with potent expectorant effects. It acts as a glucocerebrosidase (GCase) chaperone, increasing GCase activity and inducing lung autophagy. This product is intended for research and analytical applications, including studies related to Parkinson's disease and neuronopathic Gaucher disease.
  • Analytical standard for research and analytical applications.
  • Active metabolite with expectorant effects.
  • Increases glucocerebrosidase activity.
  • Induces lung autophagy.
  • Potential for Parkinson disease research.
  • Potential for neuronopathic Gaucher disease research.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-12440 Supplier Medchemexpress LLC Supplier No. HYB1039AR100MG
Ambroxol hydrochloride (Standard) is an analytical standard and an active metabolite of Bromhexine with potent expectorant effects. It acts as a glucocerebrosidase (GCase) chaperone, increasing GCase activity and inducing lung autophagy. This product is intended for research and analytical applications, including studies related to Parkinson's disease and neuronopathic Gaucher disease.
  • Analytical standard for research and analytical applications.
  • Active metabolite with expectorant effects.
  • Increases glucocerebrosidase activity.
  • Induces lung autophagy.
  • Potential for Parkinson disease research.
  • Potential for neuronopathic Gaucher disease research.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments in HPLC, GC and MS.