Learn More
Cayman Chemical Ambroxol (hydrochloride), Purity: ≥98%, CAS: 23828-92-4, 50 mg
SDP
Catalog No. 502983287
Encompass_Preferred
An expectorant; decreases short-circuit currents in electrically stimulated isolated canine tracheal epithelial cells when applied submucosally (IC50 = 60 nM); decreases IL-13-induced production of MUC5AC in primary human airway epithelial cells and inhibits IL-13-induced decreases in the mucociliary transport rate of endogenous particles in the same cells; increases phenol red secretion in mice at 30, 60, or 120 mg/kg
Catalog No. 50-298-3287 Supplier Cayman Chemical Supplier No. 1937750 MG
An expectorant; decreases short-circuit currents in electrically stimulated isolated canine tracheal epithelial cells when applied submucosally (IC50 = 60 nM); decreases IL-13-induced production of MUC5AC in primary human airway epithelial cells and inhibits IL-13-induced decreases in the mucociliary transport rate of endogenous particles in the same cells; increases phenol red secretion in mice at 30, 60, or 120 mg/kg