Learn More
AdipoGen Ambuic acid (250 µg)
SDP
Supplier:  AdipoGen AGCN20129C250
Encompass_Preferred
Chemical. CAS: 340774-69-8. Formula: C19H26O6. MW: 350.4. Isolated from Pestalotiopsis sp. Antibiotic. Antibacterial Antifungal. Inhibits the biosynthesis of cyclic peptide quormones in Gram-positive bacteria. Lead compound for antipathogenic drugs that target the quorum-sensing-mediated virulence expression of Gram-positive bacteria.
Catalog No. 50-205-3502