Learn More
Medchemexpress LLC AMC-01 1mg | 1047978-71-1 | 1 MG

Supplier: Medchemexpress LLC HY1145681MG
AMC-01 is a small-molecule chemical probe that modulates eIF2-α activity, induces PKR and NF-κB signaling, and can inhibit apoptosis. It is supplied as a white to off-white solid with high reported purity.
- Modulates eIF2-α activity and induces PKR and NF-κB.
- Inhibits apoptosis in cellular assays.
- Molecular weight 555.42 g/mol and formula C27H27BrN2O6.
- Purity 99.4% and supplied as a white to off-white solid.
- Recommended storage: -20°C sealed; in solvent: -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.