Learn More
Aat Bioquest AMC [7-Amino-4-methylcoumarin] [CAS 26093-31-2]
SDP
Catalog No. 502979201
Encompass_Preferred
This compound is the precursor for a variety of AMC enzyme substrates.
Catalog No. 50-297-9201 Supplier Aat Bioquest Supplier No. 52
This compound is the precursor for a variety of AMC enzyme substrates.