Learn More
Cayman Chemical AMC Arachidonoyl Amide, Purity: ≥98%, CAS: 862913-13-1, 50 mg
SDP
Catalog No. 502891161
Encompass_Preferred
A fluorometric FAAH substrate used for enzyme activity measurement; release of the fluorescent AMC (ex/em = 360/465 nm) allows for fast and convenient measurement of FAAH activity
Catalog No. 50-289-1161 Supplier Cayman Chemical Supplier No. 1000509850 MG
A fluorometric FAAH substrate used for enzyme activity measurement; release of the fluorescent AMC (ex/em = 360/465 nm) allows for fast and convenient measurement of FAAH activity