Learn More
Cayman Chemical AMC Arachidonoyl Amide
SDP
Catalog No. 502891229
Encompass_Preferred
A fluorometric FAAH substrate used for enzyme activity measurement; release of the fluorescent AMC (ex/em = 360/465 nm) allows for fast and convenient measurement of FAAH activity
Catalog No. 50-289-1229 Supplier Cayman Chemical Supplier No. 100050985 MG
A fluorometric FAAH substrate used for enzyme activity measurement; release of the fluorescent AMC (ex/em = 360/465 nm) allows for fast and convenient measurement of FAAH activity