Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amc arachidonoyl amide | 862913-13-1 | 99.5% | 1 MG
SDP

Catalog No. 5000232725
Encompass_Preferred

AMC Arachidonoyl Amide (AMC-AA) is a fluorogenic fatty acid amide substrate used to measure fatty acid amide hydrolase (FAAH) activity in biochemical assays. It is supplied as a light yellow to yellow solid with high purity for enzymatic studies, and should be stored at -80°C and shipped on dry ice.

  • Fluorogenic substrate for FAAH activity assays
  • High purity suitable for biochemical studies
  • Molecular weight 461.64 and formula C30H39NO3
  • Light yellow to yellow solid appearance
  • Recommended storage at -80°C and shipped on dry ice
  • Available in small quantities for assay development, such as 1 MG

Catalog No. 50-002-32725 Supplier Medchemexpress LLC Supplier No. HY1209571MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMC Arachidonoyl Amide (AMC-AA) is a fluorogenic fatty acid amide substrate used to measure fatty acid amide hydrolase (FAAH) activity in biochemical assays. It is supplied as a light yellow to yellow solid with high purity for enzymatic studies, and should be stored at -80°C and shipped on dry ice.

  • Fluorogenic substrate for FAAH activity assays
  • High purity suitable for biochemical studies
  • Molecular weight 461.64 and formula C30H39NO3
  • Light yellow to yellow solid appearance
  • Recommended storage at -80°C and shipped on dry ice
  • Available in small quantities for assay development, such as 1 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.