Learn More
AdipoGen AMCA-H
SDP
Supplier:  AdipoGen CDXA0009M500
Encompass_Preferred
Chemical. CAS 106562-32-7. Formula C12H11NO4. MW 233.22. Synthetic. AMCA is one of the brightest amine-reactive blue fluorescent dyes useful for immunofluorescence and fluorescent labeling Ex/Em 353/455nm. It is quite photostable, and its fluorescence is pH-independent from pH 4 to 10. The properties include a relatively large Stokes shift and resistance to photobleaching. Reactive AMCA derivatives are used as contrasting probes for double and triple labeling in immunofluorescence microscopy, arrays and in situ hybridization. NHS-AMCA and Sulfo-NHS-AMCA are reactive towards primary amine groups on antibodies, proteins, peptides and other biomolecules. AMCA-Hydrazide is used to label glycosylation sites or for conjugation to carboxyl groups using the crosslinker EDC.
Catalog No. 50-256-1873