Learn More
AdipoGen AMCA-H N-succinimidyl ester
SDP
Catalog No. 502561871
Encompass_Preferred
Chemical. CAS 113721-87-2. Formula C16H14N2O6. MW 330.29. Synthetic. AMCA-H NHS ester reacts with primary amine groups on antibodies, proteins, peptides and other biomolecules at pH 7.0-9.0. Suitable for labeling lysyl residues in proteins Ex/Em 353/455nm.
Catalog No. 50-256-1871 Supplier AdipoGen Supplier No. CDXA0010M100
Chemical. CAS 113721-87-2. Formula C16H14N2O6. MW 330.29. Synthetic. AMCA-H NHS ester reacts with primary amine groups on antibodies, proteins, peptides and other biomolecules at pH 7.0-9.0. Suitable for labeling lysyl residues in proteins Ex/Em 353/455nm.