Learn More
Cayman Chemical AMCA NsuccInImIdyl ester 500ug
SDP
Catalog No. 502950712
Encompass_Preferred
An amine-reactive fluorescent protein-labeling agent; abs/em = UV/400-460 nm, respectively; contains an N-hydroxysuccinimide ester that reacts with lysine residues to form photostable amide links; has commonly been used in multiplex immunophenotyping applications
Catalog No. 50-295-0712 Supplier Cayman Chemical Supplier No. 34668500 UG
An amine-reactive fluorescent protein-labeling agent; abs/em = UV/400-460 nm, respectively; contains an N-hydroxysuccinimide ester that reacts with lysine residues to form photostable amide links; has commonly been used in multiplex immunophenotyping applications