Learn More
AdipoGen AMCA-X N-succinimidyl ester
SDP
Supplier:  AdipoGen CDXA0074M010
Encompass_Preferred
Chemical. CAS 216309-02-3. Formula C22H25N3O7. MW 443.45. Synthetic. AMCA-X SE is an amine-reactive, UV-excitable, blue fluorescent dye.
Catalog No. 50-256-1838