Learn More
Biotium AMCA-X, SE
Supplier:  Biotium 90077
Encompass_Preferred
AMCA-X SE is an amine-reactive, UV-excitable, blue fluorescent dye. Properties: Ex/ Em(MeOH) = 353/442 nm; White solid soluble in DMSO or DMF; Store at -20°C and protect from light; C22H25N3O7; MW: 443.45.
Catalog No. 50-196-4820