Learn More
Selleck Chemical LLC Amcasertib (BBI503) 2mg 1129403-56-0
SDP
Supplier:  Selleck Chemical LLC S85722MG
Encompass_Preferred
Amcasertib (BBI503), a first-in-class cancer stemness kinase inhibitor, is claimed to inhibit Nanog and other CSC pathways by targeting kinases with potential anticancer activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8187