Learn More
Medchemexpress LLC Amcinonide | 51022-69-6 | 99.1% | 100 MG
SDP
Catalog No. 501970144
Encompass_Preferred
Amcinonide is an inhibitor of NO release with an IC50 of 3.38 nM. It inhibits NNC-induced expression of proinflammatory genes such as iNOS, TNF-α, and IL-1β in glial cells. It reduces the numbers of T6+/Ia+ cells, concurrently increasing T6+/Ia- cells, and induces a selective reduction in the expression of Ia antigens. Amcinonide can also be studied in research for eczematous dermatitis.
  • It inhibits nitric oxide release.
  • It inhibits the expression of proinflammatory genes in glial cells.
  • It reduces specific cell populations while increasing others.
  • It causes a selective reduction in Ia antigens.
  • It is suitable for research on eczematous dermatitis.
Catalog No. 50-197-0144 Supplier Medchemexpress LLC Supplier No. HYB1197100MG
Amcinonide is an inhibitor of NO release with an IC50 of 3.38 nM. It inhibits NNC-induced expression of proinflammatory genes such as iNOS, TNF-α, and IL-1β in glial cells. It reduces the numbers of T6+/Ia+ cells, concurrently increasing T6+/Ia- cells, and induces a selective reduction in the expression of Ia antigens. Amcinonide can also be studied in research for eczematous dermatitis.
  • It inhibits nitric oxide release.
  • It inhibits the expression of proinflammatory genes in glial cells.
  • It reduces specific cell populations while increasing others.
  • It causes a selective reduction in Ia antigens.
  • It is suitable for research on eczematous dermatitis.