Learn More
TARGETMOL CHEMICALS INC Amcinonide 200MG
SDP
Catalog No. 502880535
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Amcinonide (CL-34699) is a Corticosteroid. The mechanism of action of amcinonide is as a Corticosteroid Hormone Receptor Agonist. Purity 99.9%
Catalog No. 50-288-0535 Supplier TARGETMOL CHEMICALS INC Supplier No. T0261200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Amcinonide (CL-34699) is a Corticosteroid. The mechanism of action of amcinonide is as a Corticosteroid Hormone Receptor Agonist. Purity 99.9%