Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ AMD-3+ Multiple Delivery 6psi 3 Flowmeter Inhalant Anesthesia System With Vaporizer
SDP

Catalog No. 04777104 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
04-777-104 Research Animal Inhalant Anesthesia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-777-104 Supplier SOMNI Scientific™ Supplier No. AMD17106
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The ultimate in Inhalant Anesthesia gas delivery. This pressurized system can deliver accurate flowrates to 3 stations simultaneously. Each station is equipped with a flowmeter and toggle switch allowing the user to leave flows preset or adjust as needed.

  • 3 Flowmeter Controlled Stations with Toggle Switches
  • Color Coded delivery lines to each station
  • Common Gas outlets for each station
  • Small footprint
  • Adjustable flowrates
  • Stacking design save valuable workbench space
  • Includes TT-2 with 6psi Calibrated vaporizer and three 6 ft. color coded delivery lines with adapters

Specifications

For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) EM-16000, EM-16001, EM-16002, EM-16003, EM-16004, EM-16005, EM-16006, EM-16007, EM-16008, EM-16010, EM-16011, IA-3009, OC-21003, WG-15001, WG-15002, WG-15003
Material Aluminum, Brass, Stainless Steel, Plastic
Length (Metric) 30.48 cm
Width (Metric) 20.32 cm
Includes 6 psi calibrated Vaporizer, TT frame, Pressure relief valve and three 6 ft. color-coded delivery lines with adapters

For Veterinary Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.