Learn More
Medchemexpress LLC HY-15971 5mg Medchemexpress, AMD 3465 (hexahydrobromide) CAS:185991-07-5 Purity:>98%
SDP
Catalog No. 502081853
Encompass_Preferred
Medchemexpress, HY-15971 5mg AMD 3465 (hexahydrobromide) CAS:185991-07-5 AMD 3465 hexahydrobromide (GENZ-644494 hexahydrobromide) is a potent antagonist of CXCR4, inhibits binding of 12G5 mAb and CXCL12AF647 to CXCR4, with IC50s of 0.75 nM and 18 nM in SupT1 cells; AMD 3465 also potently inhibits the replication of X4 HIV strains (IC50: 1-10 nM), but has no effect on CCR5-using (R5) viruses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1853 Supplier Medchemexpress LLC Supplier No. HY159715MG
Medchemexpress, HY-15971 5mg AMD 3465 (hexahydrobromide) CAS:185991-07-5 AMD 3465 hexahydrobromide (GENZ-644494 hexahydrobromide) is a potent antagonist of CXCR4, inhibits binding of 12G5 mAb and CXCL12AF647 to CXCR4, with IC50s of 0.75 nM and 18 nM in SupT1 cells; AMD 3465 also potently inhibits the replication of X4 HIV strains (IC50: 1-10 nM), but has no effect on CCR5-using (R5) viruses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.