Learn More
Abcam AMD 3465 he x ahydrobromide, C x CR4 antagonist, 10MG
Catalog No. 502707242
Encompass_Preferred
MW 410.6 Da, Purity >97%. Potent and selective CXCR4 antagonist. Shows 8-fold greater affinity than AMD 3100 (ab120718). Potent HIV entry inhibitor (IC₅₀ = ~10 nM). Additionally mobilizes stem cells in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7242 Supplier Abcam Supplier No. AB12080910MG
MW 410.6 Da, Purity >97%. Potent and selective CXCR4 antagonist. Shows 8-fold greater affinity than AMD 3100 (ab120718). Potent HIV entry inhibitor (IC₅₀ = ~10 nM). Additionally mobilizes stem cells in vivo.
The product is subject to the following: Abcam Restricted Use Statement