Learn More
Cayman Chemical AMD 3465hydrobromIde 25mg
SDP
Catalog No. 502955767
Encompass_Preferred
Blocks the cell surface binding of the CXCR4 ligand CXCL12 (IC50 = 18 nM), inhibits calcium mobilization (IC50 = 4 nM), and (at 6,250 nM) completely blocks chemotaxis of SupT1 cells; active against various HIV strains (IC50s = 6-12 nM)
Catalog No. 50-295-5767 Supplier Cayman Chemical Supplier No. 1195925 MG
Blocks the cell surface binding of the CXCR4 ligand CXCL12 (IC50 = 18 nM), inhibits calcium mobilization (IC50 = 4 nM), and (at 6,250 nM) completely blocks chemotaxis of SupT1 cells; active against various HIV strains (IC50s = 6-12 nM)