Learn More
Electron Microscopy Sciences AMD-SA Balance Table With Isolation Area, 75 x 75 cm, Stainless Steel
SDP
Catalog No. 5034476
Encompass_Preferred
This stainless air table features our AMB-30x45 pneumatic platform. It incorporates four low 2Hz natural frequency rolling diaphragm damped isolators and the platform and table top feature 240 ground satin stainless steel. The isolators are simply inflated via adjustable valves using a bicycle pump. Adjustable valves using a bicycle pump.
Size: 75 x 75 cm
Type: Stainless Steel
Surface: Passive Air Pump Up
Catalog No. 50-344-76 Supplier Electron Microscopy Sciences Supplier No. 6705
This stainless air table features our AMB-30x45 pneumatic platform. It incorporates four low 2Hz natural frequency rolling diaphragm damped isolators and the platform and table top feature 240 ground satin stainless steel. The isolators are simply inflated via adjustable valves using a bicycle pump. Adjustable valves using a bicycle pump.
Size: 75 x 75 cm
Type: Stainless Steel
Surface: Passive Air Pump Up