Learn More
Cole-Parmer Speirs Robertson AMD-SS-75x75 AMD-SS Balance Table, 75x75 cmwith 30x45 cmIsolation Platform
Catalog No. NC1650011 Shop All Cole-Parmer Products
Encompass
These worktables eliminate unwanted vibrations which limit the performance of sensitive instruments such as balances. Both series feature a sound-deadened isolation platform core and tubular steel frame with leveling feet. The isolation platform can be removed for cleaning, or switched with other platforms as necessary.
This item is not returnable. View return policy
Catalog No. NC1650011 Supplier Cole-Parmer Supplier No. 1150046
These worktables eliminate unwanted vibrations which limit the performance of sensitive instruments such as balances. Both series feature a sound-deadened isolation platform core and tubular steel frame with leveling feet. The isolation platform can be removed for cleaning, or switched with other platforms as necessary.