Learn More
Aviva Systems Biology Corp AMD1 Antibody - N-terminal region (ARP54480_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54480_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AMD1. Alternate names: ADOMETDC, AMD, DKFZp313L1234, FLJ26964, SAMDCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61218