Learn More
Sigma Aldrich Fine Chemicals Biosciences AMD3100 octahydrochloride hydrate, 155148-31-5 (anhydrous), MFCD00927130, 5mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A56025MG
Encompass
Molecular Formula: C28H54N8 · 8HCl · xH2O, Molecular Weight: 794.47 (anhydrous basis), ≥97% (NMR), solid
Catalog No. NC1026659