Learn More
Aviva Systems Biology Corp AMDHD2 Antibody - N-terminal region (ARP63138_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63138_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AMDHD2. It was validated on Western Blot. . Alternate names: CGI-14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-892