Learn More
Medchemexpress LLC Amdiglurax phosphate | 1270138-41-4 | 99.8% | 464.49 g·mol⁻¹ | C22H33N4O5P | 5 MG
SDP
Catalog No. 5000222213
Encompass_Preferred
Amdiglurax phosphate is the phosphate salt of Amdiglurax (NSI-189), a small-molecule research compound reported to promote neurogenesis and synaptic plasticity. It has been investigated for neuroprotective, antidepressant, and procognitive effects and is supplied for laboratory research use.
  • Phosphate salt of a neurogenic small molecule.
  • Reported to enhance neurogenesis and synaptic plasticity.
  • Demonstrated up-regulation of neurogenic factors such as BDNF and SCF.
  • Investigated for antidepressant and procognitive effects.
  • Provided with high purity suitable for research applications.
  • Soluble in DMSO for experimental use.
Catalog No. 50-002-22213 Supplier Medchemexpress LLC Supplier No. HY13999A15MG
Amdiglurax phosphate is the phosphate salt of Amdiglurax (NSI-189), a small-molecule research compound reported to promote neurogenesis and synaptic plasticity. It has been investigated for neuroprotective, antidepressant, and procognitive effects and is supplied for laboratory research use.
  • Phosphate salt of a neurogenic small molecule.
  • Reported to enhance neurogenesis and synaptic plasticity.
  • Demonstrated up-regulation of neurogenic factors such as BDNF and SCF.
  • Investigated for antidepressant and procognitive effects.
  • Provided with high purity suitable for research applications.
  • Soluble in DMSO for experimental use.