Learn More
Research Products International Corp Amdinocillin [Mecillinam], 1 Gram
SDP
Catalog No. 501368099
Encompass_Preferred
Amdinocillin is a broad-spectrum antibiotic that belongs to the penicillin class of antibiotics. It inhibits the synthesis of the bacterial cell wall, leading to the destruction of the pathogen and is effective against both gram-positive and gram-negative bacteria.
Catalog No. 50-136-8099 Supplier Research Products International Corp Supplier No. A916901.0
Amdinocillin is a broad-spectrum antibiotic that belongs to the penicillin class of antibiotics. It inhibits the synthesis of the bacterial cell wall, leading to the destruction of the pathogen and is effective against both gram-positive and gram-negative bacteria.