Learn More
Alkali Scientific Amdinocillin [Mecillinam], 10 G
SDP
Catalog No. 5000661092
Encompass_Preferred
Amdinocillin, also known as mecillinam, is a beta lactam antibiotic used for its activity against Gram negative bacteria. It works by inhibiting bacterial cell wall synthesis through binding to specific penicillin binding proteins, ultimately leading to cell lysis and death. This antibiotic is widely used in microbiology research for studying bacterial growth, resistance mechanisms, and antimicrobial susceptibility. Its selective activity makes it particularly useful for targeting Enterobacteriaceae species. In laboratory applications, it is used in culture media and experimental setups requiring controlled inhibition of bacterial populations, supporting research in microbiology, pharmacology, and antibiotic development.
Catalog No. 50-006-61092 Supplier Alkali Scientific Supplier No. AMD001CP
Add to Cart
Add to Cart
Amdinocillin, also known as mecillinam, is a beta lactam antibiotic used for its activity against Gram negative bacteria. It works by inhibiting bacterial cell wall synthesis through binding to specific penicillin binding proteins, ultimately leading to cell lysis and death. This antibiotic is widely used in microbiology research for studying bacterial growth, resistance mechanisms, and antimicrobial susceptibility. Its selective activity makes it particularly useful for targeting Enterobacteriaceae species. In laboratory applications, it is used in culture media and experimental setups requiring controlled inhibition of bacterial populations, supporting research in microbiology, pharmacology, and antibiotic development.