Learn More
Alkali Scientific Amdinocillin [Mecillinam], 25 G
SDP
Supplier:  Alkali Scientific AMD002CP
Encompass_Preferred
Amdinocillin, also known as mecillinam, is a beta lactam antibiotic effective against a range of Gram negative bacteria. It functions by disrupting bacterial cell wall synthesis through selective binding to penicillin binding proteins, leading to structural instability and cell death. This compound is widely used in microbiological research to study antimicrobial activity, bacterial physiology, and resistance mechanisms. Its targeted spectrum makes it especially useful for working with Enterobacteriaceae species in laboratory settings. The product is commonly incorporated into culture media or experimental assays where controlled bacterial inhibition is required, supporting research in microbiology, biotechnology, and drug development.
Catalog No. 50-006-60807