Learn More
Teknova amdS Biolistic Agar Plates. 100mm, 20 Plates per Sleeve, Sterile 4 pack.
SDP
Catalog No. 502387761
Encompass
2.00% Noble Agar, 2.00% Glucose, 1.50% Monopotassium Phosphate,0.17% Cesium Chloride, 0.06% Acetamide, 0.06% Calcium Chloride, 0.06% Magnesium Sulfate,5.0g/mL Ferrous Sulfate, 1.6g/mL Manganese Sulfate, 1.4g/mL Zinc Sulfate, 1.0g/mL Cobalt Chloride
Catalog No. 50-238-7761 Supplier Teknova Supplier No. V01684PK
2.00% Noble Agar, 2.00% Glucose, 1.50% Monopotassium Phosphate,0.17% Cesium Chloride, 0.06% Acetamide, 0.06% Calcium Chloride, 0.06% Magnesium Sulfate,5.0g/mL Ferrous Sulfate, 1.6g/mL Manganese Sulfate, 1.4g/mL Zinc Sulfate, 1.0g/mL Cobalt Chloride