Learn More
Accuflow Systems AME-CLAMP PINCH CLAMP 50 per Pack
Catalog No. 501109800
Encompass_Preferred
WL GORE Clamp,Ame Clamp, WL GORE/Amesil Clamp,uniquely designed closure for tubing or hose that requires Higher clamp pressures. Can be utilized with a variety of tubing and hose sizes. Quckly installed with a positive swingaway locking screw. Sanitary , Reusable, Lightweight , Heavy duty high strength polymer construction, PK/50 ea
Catalog No. 50-110-9800 Supplier Accuflow Systems Supplier No. AMS4232PK50
WL GORE Clamp,Ame Clamp, WL GORE/Amesil Clamp,uniquely designed closure for tubing or hose that requires Higher clamp pressures. Can be utilized with a variety of tubing and hose sizes. Quckly installed with a positive swingaway locking screw. Sanitary , Reusable, Lightweight , Heavy duty high strength polymer construction, PK/50 ea