Learn More
Accuflow Systems W. L. Gore Tube Clamp, Polysulfone, Pinch Closure, Autoclavable
Catalog No. NC3666684
Encompass_Preferred
W. L. Gore AME-Clamp is a high-strength polysulfone tube clamp designed for sanitary and bioprocess tubing applications. It provides a secure pinch closure for tubing and hose, installs quickly with a swingaway locking screw, and is reusable and cleanable for repeated sterilization cycles.
  • Polysulfone construction for chemical resistance and durability.
  • Autoclavable up to 30 cycles, gamma compatible up to 50 kGy.
  • FDA-compliant material suitable for bioprocess environments.
  • Compatible with unreinforced tubing wall thickness .090-.190 in.
  • Compatible with reinforced braided hose wall thickness .090-.180 in.
  • Positive swingaway locking screw for quick, tool-free installation.
  • Reusable and fully disassemblable for cleaning and maintenance.
  • Suitable for silicone, C-Flex, PVC, and similar elastomeric tubing.
Catalog No. NC3666684 Supplier Accuflow Systems Supplier No. AMS4232EA
W. L. Gore AME-Clamp is a high-strength polysulfone tube clamp designed for sanitary and bioprocess tubing applications. It provides a secure pinch closure for tubing and hose, installs quickly with a swingaway locking screw, and is reusable and cleanable for repeated sterilization cycles.
  • Polysulfone construction for chemical resistance and durability.
  • Autoclavable up to 30 cycles, gamma compatible up to 50 kGy.
  • FDA-compliant material suitable for bioprocess environments.
  • Compatible with unreinforced tubing wall thickness .090-.190 in.
  • Compatible with reinforced braided hose wall thickness .090-.180 in.
  • Positive swingaway locking screw for quick, tool-free installation.
  • Reusable and fully disassemblable for cleaning and maintenance.
  • Suitable for silicone, C-Flex, PVC, and similar elastomeric tubing.