Learn More
MSC AME International Forklift Jack, 7.5 ton, 2.25 in Low Clearance
Catalog No. 5000515155
Encompass
This forklift jack is designed for high-capacity lifting applications. It features a 7.5 ton capacity and a low clearance, making it suitable for various industrial and automotive tasks. The jack provides a substantial lifting height for efficient maintenance and repair operations.
  • Low clearance of 2.25 in (5.7 cm)
  • Maximum lifting height of 16.6 in (41 cm)
  • 7.5 ton capacity
  • 99 lb (45 kg) weight
Catalog No. 50-005-15155 Supplier MSC Supplier No. 95656039
This forklift jack is designed for high-capacity lifting applications. It features a 7.5 ton capacity and a low clearance, making it suitable for various industrial and automotive tasks. The jack provides a substantial lifting height for efficient maintenance and repair operations.
  • Low clearance of 2.25 in (5.7 cm)
  • Maximum lifting height of 16.6 in (41 cm)
  • 7.5 ton capacity
  • 99 lb (45 kg) weight