Learn More
Medchemexpress LLC Amelparib hydrochloride | 98.4% | 379.88 | 1 MG
SDP
Catalog No. 5000436206
Encompass_Preferred
Amelparib hydrochloride is a potent, orally active, and water-soluble inhibitor of PARP-1. It inhibits PARP-1 activity (IC50 = 18.5 nM) and cellular PAR formation (IC50 = 10.7 nM). This compound is a potential neuroprotective agent and has potential for the research of acute ischaemic stroke.
  • Potent, orally active, and water-soluble inhibitor of PARP-1
  • Inhibits PARP-1 activity (IC50 = 18.5 nM)
  • Inhibits cellular PAR formation (IC50 = 10.7 nM)
  • Potential neuroprotective agent
  • Potential for research of acute ischaemic stroke
  • Appearance: Solid
  • Color: Off-white to light yellow
Catalog No. 50-004-36206 Supplier Medchemexpress LLC Supplier No. HY116218C1MG
Amelparib hydrochloride is a potent, orally active, and water-soluble inhibitor of PARP-1. It inhibits PARP-1 activity (IC50 = 18.5 nM) and cellular PAR formation (IC50 = 10.7 nM). This compound is a potential neuroprotective agent and has potential for the research of acute ischaemic stroke.
  • Potent, orally active, and water-soluble inhibitor of PARP-1
  • Inhibits PARP-1 activity (IC50 = 18.5 nM)
  • Inhibits cellular PAR formation (IC50 = 10.7 nM)
  • Potential neuroprotective agent
  • Potential for research of acute ischaemic stroke
  • Appearance: Solid
  • Color: Off-white to light yellow